Sodium dihydrogen phosphate 3 M

Supplier: Rigaku Reagents

Synonyms: Sodium dihydrogenphosphate (granular), Sodium dihydrogen orthophosphate, Monobasic Sodium Phosphate, Sodium phosphate monobasic, Sodium dihydrogen phosphate

1008372 1008373
101443-396EA 102 USD
101443-396 101443-756
Sodium dihydrogen phosphate 3 M
Sodium dihydrogen phosphate

Use our stock solutions directly or as a component in solutions you create in your own lab. We prepare our stock solutions using high quality raw materials, ASTM Type 1 water, and sterile packaging. Our stock solutions are intended for Research and Development Use Only.

Formula: NaH₂PO₄
MDL Number: MFCD00003527
CAS Number: 7558-80-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR