Disodium-L(+)-tartrate solution 1 M
Supplier: Rigaku Reagents
1008282
1008283
101443-416EA
92.85
USD
101443-416
101443-806
Disodium-L(+)-tartrate solution 1 M
Disodium-L(+)-tartrate solutions
Use our stock solutions directly or as a component in solutions you create in your own lab. We prepare our stock solutions using high quality raw materials, ASTM Type 1 water, and sterile packaging. Our stock solutions are intended for Research and Development Use Only.
Formula:
NaOOC(CHOH)₂COONa·2H₂O MW: 230.08 g/mol |
MDL Number:
MFCD00150035 CAS Number: 6106-24-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...