Disodium malonate 2 M
Supplier: Rigaku Reagents
Synonyms:
Malonic acid disodium salt
, Sodium malonate dibasic
1008345
101443-782EA
149.08
USD
101443-782
Disodium malonate 2 M
Disodium malonate
Use our stock solutions directly or as a component in solutions you create in your own lab. We prepare our stock solutions using high quality raw materials, ASTM Type 1 water, and sterile packaging. Our stock solutions are intended for Research and Development Use Only.
Formula:
C₃H₂Na₂O₄ |
MDL Number:
MFCD00002708 CAS Number: 141-95-7 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...