Sodium iodide 1 M
Supplier: Rigaku Reagents
Synonyms:
Ioduril
, Hydriodic acid sodium salt
, Sodium monoiodide
1008267
1008266
101443-760EA
126.85
USD
101443-760
101443-398
Sodium iodide 1 M
Sodium iodide
Use our stock solutions directly or as a component in solutions you create in your own lab. We prepare our stock solutions using high quality raw materials, ASTM Type 1 water, and sterile packaging. Our stock solutions are intended for Research and Development Use Only.
Formula:
NaI MW: 149.89 g/mol |
MDL Number:
MFCD00003532 CAS Number: 7681-82-5 UN: 3077 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...