Sodium fluoride 0.5 M
Supplier: Rigaku Reagents
1008046
1008047
101443-390EA
117.69
USD
101443-390
101443-750
Sodium fluoride 0.5 M
Sodium fluoride
Use our stock solutions directly or as a component in solutions you create in your own lab. We prepare our stock solutions using high quality raw materials, ASTM Type 1 water, and sterile packaging. Our stock solutions are intended for Research and Development Use Only.
Formula:
NaF |
MDL Number:
MFCD00003524 CAS Number: 7681-49-4 UN: 1690 ADR: 6.1,III |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...