tri-Sodium citrate solution 1.6 M
Supplier: Rigaku Reagents
1008082
1008083
101443-386EA
82.38
USD
101443-386
101443-744
tri-Sodium citrate solution 1.6 M
tri-Sodium citrate solutions
Use our stock solutions directly or as a component in solutions you create in your own lab. We prepare our stock solutions using high quality raw materials, ASTM Type 1 water, and sterile packaging. Our stock solutions are intended for Research and Development Use Only.
Formula:
Na₃C₆H₅O₇·2H₂O MW: 294.1 g/mol |
MDL Number:
MFCD00150031 CAS Number: 6132-04-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...