Potassium acetate 1 M
Supplier: Rigaku Reagents
Synonyms:
4-Chloro-N-furfuryl-5-sulfamoylanthranlic acid, Diuretic salt, Acetic acid potassium salt, Acetic acid potassium salt
1008194
1008195
101443-316EA
81.08
USD
101443-316
101443-664
Potassium acetate 1 M
Potassium acetate
Use our stock solutions directly or as a component in solutions you create in your own lab. We prepare our stock solutions using high quality raw materials, ASTM Type 1 water, and sterile packaging. Our stock solutions are intended for Research and Development Use Only.
Formula:
CH₃COOK MW: 98.14 g/mol Melting Pt: 304 |
MDL Number:
MFCD00012458 CAS Number: 127-08-2 Merck Index: 13,07687 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...