Nickel(II) chloride 4 M

Supplier: Rigaku Reagents

1008380
101443-448EA 173.92 USD
101443-448
Nickel(II) chloride 4 M
Nickel(II) chloride solutions

Use our stock solutions directly or as a component in solutions you create in your own lab. We prepare our stock solutions using high quality raw materials, ASTM Type 1 water, and sterile packaging. Our stock solutions are intended for Research and Development Use Only.

Formula: NiCl₂·6H₂O
MW: 237.69 g/mol
MDL Number: MFCD00149809
CAS Number: 7791-20-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR