Magnesium sulfate 2 M

Supplier: Rigaku Reagents

1008340 1008341
101443-378EA 96.77 USD
101443-378 101443-736
Magnesium sulfate 2 M
Magnesium sulphate solutions

Use our stock solutions directly or as a component in solutions you create in your own lab. We prepare our stock solutions using high quality raw materials, ASTM Type 1 water, and sterile packaging. Our stock solutions are intended for Research and Development Use Only.

Formula: MgSO₄·7H₂O
MDL Number: MFCD00149785
CAS Number: 10034-99-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR