Ammonium sulfate 3.5 M

Supplier: Rigaku Reagents

Synonyms: sulfuric acid diammonium salt, Diammonium sulfate, Sulphuric acid diammonium salt, Diammonium sulphate, Mascagnite

1008358 1008359
101443-442EA 86.31 USD
101443-442 101443-834
Ammonium sulfate 3.5 M
Ammonium sulphate

Use our stock solutions directly or as a component in solutions you create in your own lab.


We prepare our stock solutions using high quality raw materials, ASTM Type 1 water, and sterile packaging. Our stock solutions are intended for Research and Development Use Only.

Formula: (NH₄)₂SO₄
MW: 132.14 g/mol
MDL Number: MFCD00003391
CAS Number: 7783-20-2
Merck Index: 13,00559

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR