Diammonium hydrogen orthophosphate 2 M

Supplier: Rigaku Reagents

Synonyms: Diammonium hydrogen orthophosphate, Ammonium hydrogen phosphate, di-Ammonium hydrogen phosphate, Ammonium Phosphate, Ammonium biphosphate, Fyrex, Ammonium phosphate dibasic, Ammonium monohydrogen phosphate, Diammonium hydrogen phosphate

1008331 1008330
101443-830EA 142.54 USD
101443-830 101443-440
Diammonium hydrogen orthophosphate 2 M
di-Ammonium hydrogen orthophosphate

Use our stock solutions directly or as a component in solutions you create in your own lab. We prepare our stock solutions using high quality raw materials, ASTM Type 1 water, and sterile packaging. Our stock solutions are intended for Research and Development Use Only.

Formula: (NH₄)₂HPO₄
MW: 132.06 g/mol
Melting Pt: 185
MDL Number: MFCD00010891
CAS Number: 7783-28-0
Merck Index: 13,00546

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR