Diammonium hydrogen citrate 1 M

Supplier: Rigaku Reagents

Synonyms: Ammonium citrate dibasicDiammonium hydrogen 2-hydroxypropane-1,2,3-tricarboxylateCitric acid diammonium salt

1008098 1008097
101443-276EA 87.62 USD
101443-276 101443-610
Diammonium hydrogen citrate 1 M
di-Ammonium hydrogen citrate

Use our stock solutions directly or as a component in solutions you create in your own lab. We prepare our stock solutions using high quality raw materials, ASTM Type 1 water, and sterile packaging. Our stock solutions are intended for Research and Development Use Only.

Formula: HOC(CO₂H)(CH₂CO₂NH₄)₂
MW: 226.19 g/mol
MDL Number: MFCD00013068
CAS Number: 3012-65-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR