(±)-2-Butanol ≥99.5%

Supplier: Rigaku Reagents

Synonyms: (±)-sec-Butanol, (±)-sec-Butyl alcohol, (±)-Ethyl methyl carbinol, (±)-2-Hydroxy butane, sec-Butanol, sec-Butyl alcohol, (±)-Methyl ethyl carbinol

1008396 1008397
101443-242EA 71.92 USD
101443-242 101443-558
(±)-2-Butanol ≥99.5%
(±)-2-Butanol

Use our stock solutions directly or as a component in solutions you create in your own lab. We prepare our stock solutions using high quality raw materials, ASTM Type 1 water, and sterile packaging. Our stock solutions are intended for Research and Development Use Only.

Formula: H₃CCH₂CH(OH)CH₃
MW: 74.12 g/mol
Boiling Pt: 94
Melting Pt: -115
Flash Pt: 23
MDL Number: MFCD00004569
CAS Number: 78-92-2
UN: 1120
ADR: 3 III
Merck Index: 13,01540
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1dea3607167631fbfcf7751c20b71f345ad24c19&vid=010e04e8f508fd3d501873d20c2349817834b6e8&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR