Proto® TorquePlus™ Impact Sockets, ORS Nasco
Supplier: Safety & Industrial Supplies
577-07523 577-07524 577-07509 577-07510T 577-07511T 577-07512 577-07515T 577-07516S 577-07516T 577-07517 577-07518S 577-07522T 577-07523T 577-07524T 577-07528 577-07526T 577-07527T 577-07532T 577-07534T 577-07536T 577-07510 577-07516 577-07521T 577-07521 577-07518 577-07520 577-07526 577-07511 577-07512T 577-07519 577-07519T 577-07525 577-07527 577-07529 577-07538 577-07532 577-07513 577-07514 577-07514S 577-07514T 577-07515 577-07517S 577-07517T 577-07518T 577-07519S 577-07520S 577-07520T 577-07521S 577-07522 577-07530 577-07533 577-07533T 577-07534 577-07535 577-07536 577-07528T 577-07529T 577-07530T
300002-676EA 48.3 USD
300002-676 300002-680 300022-778 300022-782 300022-790 300022-798 300022-800 300022-842 300022-850 300022-852 300022-854 300022-868 300022-920 300022-928 300022-940 300022-964 300030-710 300030-712 300030-722 300030-724 300030-726 300030-728 300022-784 300022-844 300022-908 300022-896 300002-670 300002-674 300002-682 300022-792 300022-808 300022-872 300022-884 300022-942 300022-956 300022-970 300023-020 300002-686 300022-810 300022-824 300022-830 300022-832 300022-834 300022-862 300022-864 300022-870 300022-882 300022-892 300022-894 300022-906 300022-910 300022-976 300022-994 300023-000 300023-002 300023-006 300023-012 300030-714 300030-716 300030-718 300030-720 300030-730
Proto® TorquePlus™ Impact Sockets, ORS Nasco
Tool Sets
The TorquePlus™ internal design minimizes fastener rounding, helps to extend fastener life, and allows the user to more easily turn worn, rounded, or rusted fasteners.
Proto® Impact Sockets are ideal for use on stubborn or damaged fasteners, and also on fasteners made from soft materials. The TorquePlus™ design is a high-performance drive configuration which allows for quick and easy socket engagement to the fastener flats, not the corners.