Microbiology Slide, Microbiologics
Supplier: MICROBIOLOGICS, INC.
SL10-10 SL15-10 SL40-10 SL41-10 SL50-10 SL60-10 SL70-10 SL75-10 SL90-10 SL45-10
89508-208EA 132.92 USD
89508-208 89508-210 89508-212 89508-214 89508-218 89508-220 89508-222 89508-224 89508-228 89508-217
Microbiology Slide, Microbiologics
Specimens Preserved Specimens Preserved Bacteria Materials
Microscopy staining is a tried and true method for examining the cellular morphology of microorganisms. Laboratory best practices should include quality control measures to ensure that staining materials and processes are producing accurate results. We offer a variety of QC Microbiology Slides containing droplets of an air-dried, fixed and preserved organism, or a smear containing a designated organism population.
  • Convenient test ready format saves you time and money
  • Room temperature storage is easy and economical
  • Technical Support experts available for guidance
Ordering information: To receive this product, customers must have a valid ATCC End User Licensing Agreement in place with Microbiologics. Registration for this agreement is only required once, per location during the first order. Instructions for how to register can be downloaded from the product page. Go to the "Related Information" box, click on "Additional Product Details", then "Instructions for End User Registration with Microbiologics".
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Microbiologics Inc. is licensed to use these trademarks and to sell products derived from ATCC® cultures.