Pentaerythritol propoxylate (17/8 PO/OH) 50% (v/v)

Supplier: Rigaku Reagents

1008071
101443-890EA 125.54 USD
101443-890
Pentaerythritol propoxylate (17/8 PO/OH) 50% (v/v)
Pentaerythritol propoxylate
Formula: C[CH₂[OCH₂CH(CH₃)]nOH]₄
MDL Number: MFCD00197885
CAS Number: 9051-49-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR