Aluminum Tuning Fork, Set with Hammer
Supplier: EISCO SCIENTIFIC LLC
PH0738BN8
470230-250EA 60 USD
470230-250
Aluminum Tuning Fork, Set with Hammer
Educational Materials Physics Educational Materials Waves and Oscillations Learning Activities
Aluminum tuning forks for physics experiments.
  • C (256), D (288), E (320), F (341), G (384), A (426), B (480), C (512)
  • Comes with mallet and wood case
  • Designed for physics experiments and actual frequencies can be ±5% stated Hz
Premium aluminum physics tuning fork set with hammer. Tuning forks included: C (256), D (288), E (320), F (341), G (384), A (426), B (480), C (512). Designed for Physics experiments and actual frequencies can be ±5% stated Hz.