Inert Arc Nipples, Western Enterprises
Supplier: Safety & Industrial Supplies
312-AW-16 312-AW-18 312-AW-19 312-AW-16B 312-AW-13 312-AW-20 312-AW-17 312-AW-3
300007-917EA 7.33 USD
300007-917 300007-919 300007-920 300066-510 300066-506 300066-512 300007-918 300066-524
Inert Arc Nipples, Western Enterprises
Welding Supplies
Made from brass to ensure functionality and durability.
Inert arc nipples can accommodate up to 200 psi of pressure. Use these arc nipples to repair or make a new hose to attach to the shielding gas (or cooling) lines.