Barbed Hose Nipples, Western Enterprises
Supplier: Safety & Industrial Supplies
312-18 312-20 312-19 312-17 312-C-18 312-20-A 312-C-17 312-D-18 312-216
300007-810EA 1.39 USD
300007-810 300007-812 300066-684 300066-686 300007-811 300067-000 300007-809 300007-949 300066-290 300066-682 300066-754 300091-792
Barbed Hose Nipples, Western Enterprises
Welding Supplies
Made from brass to ensure functionality and durability
Can accommodate up to 200 psi of pressure.