Knife Switches
Supplier: Wards
PH1000A PH1000B PH1002A PH1002B
470230-366EA 7.8 USD
470230-366 470230-368 470230-370 470230-372
Knife Switches
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Durable plastic knife switch for electrical circuits.
  • Single Pole, single throw knife switch
  • Durable plastic base for student use
  • Brass poles and knives for strong electrical connection
Single pole, single throw knife switch on a durable plastic base. Rugged construction that stands up to tough student use. Poles and knives are made of brass for good electrical connections.