SP Wilmad-LabGlass Precision PTFE Heavy Duty Stirrer Bearing Assemblies, SP Industries
Supplier: SP Industries
LG-9450-102 LG-9450-148 LG-9450-146 LG-9450-100 LG-9450-140 LG-9450-112 LG-9450-142 LG-9450-114 LG-9450-144
14201-590EA 36.34 USD
14201-590 14203-922 14207-042 14203-858 14201-650 14212-348 14202-388 14207-126 14206-486
SP Wilmad-LabGlass Precision PTFE Heavy Duty Stirrer Bearing Assemblies, SP Industries
Stirrers Stirring Tools
These uniquely designed, rugged, heavy duty stirrer bearings are used under heavier torque conditions.
Complete assembly includes PTFE stirrer bearing, PTFE bushing, saddle O-ring, Viton® O-ring, threaded heavy wall glass adapter, and top screw cap. Use a 19 mm stirrer shaft (LG-9504) with this bearing.
The 19 mm I.D., self-aligning, PTFE bearings and saddle O-rings provide a constant positive seal on the stirring rod.
Ordering information: Ground finish rods from other manufacturers are not recommended for use with this bearing.