Tryptic Soy Broth, Tubed Culture Media, bioMerieux
Supplier: bioMerieux
42100 P1720
95060-764CS 56.41 USD
95060-764 95061-570 97067-340
Tryptic Soy Broth, Tubed Culture Media, bioMerieux
Media for Microbiology Prepared Media for Microbiology
Tryptic Soy Broth is a general-purpose medium used in qualitative procedures for the cultivation of fastidious and nonfastidious microorganisms.
  • General purpose medium
  • Multiple sizes available
General purpose medium for cultivation of a wide variety of microorganisms recommended by USP/EP/JP for sterility testing of pharmaceutical article
Store according to the package label in an upright position in the dark. Do not freeze. Allow warming of medium to room temperature before inoculation.
The medium contains two peptones that are excellent sources of nitrogen, amino acids, and peptides. It supports the growth of a wide variety of microorganisms. Common aerobic and facultative anaerobic bacteria grow readily.