Mueller Hinton Broth, HiMedia Laboratories
Supplier: HiMedia
M391-2.5KG M391-5KG
89225-394EA 741.69 USD
89225-394 89225-396
Mueller Hinton Broth, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Used to determine the susceptibility of bacteria to Sulphonamides by the tube dilution method.
  • Multiple sizes available