SpillFix Granular Absorbent, Brady Worldwide
Supplier: Avantor
149063 149116
76020-708EA 61.63 USD
76020-708 76020-718 76020-720 76020-719
SpillFix Granular Absorbent, Brady Worldwide
Spill Control Materials
SpillFix granular is completely natural, producing no carcinogens or dust
  • Premium absorbent made of 100% recycled material
  • Extremely effective on the absorption of hydrocarbons, water-based fluids, and most aggressive chemicals
  • Certified safe for landfill disposal
  • Lightweight and easy to deploy
  • Convenient 3 quart jar is perfect for smaller spills and tight spaces
It has more than four times the absorption power of clay granular.