VWR® First Aid Metal Cabinets, ANSI A+, Type I & II, with Medications
Supplier: VWR International
76049-056 76049-060
76049-056CS 621.5 USD
76049-056 76049-060
VWR® First Aid Metal Cabinets, ANSI A+, Type I & II, with Medications
First Aid Supplies First Aid Cabinets
These steel first aid cabinets meet ANSI standards and are designed for worksites and companies with 75+ employees.
  • Solid steel case, with carrying handle
  • Wall-mountable
  • Available in a 2-shelf or 3-shelf configuation
  • Available with 347 or 669 pieces
This solid steel case comes with a carrying handle and is wall-mountable.
Certifications: ANSI/ISEA Z308.1-2015. The 2-shelf cabinet exceeds the 2015 Class B requirements, and is sufficient to treat 75 to 100 people. The 3-shelf cabinet exceeds the 2015 Class A requirements, and is sufficient to treat 10More Product Information