Ear and Eye Protection Area Sign, Brady Worldwide
Supplier: Brady
25194 88543 42772 42771 88544 25195
76046-552EA 40.11 USD
76046-552 76046-504 76046-506 76046-482 76046-550 76046-484
Ear and Eye Protection Area Sign, Brady Worldwide
Signs
Personal Protection (PPE) signs.
  • B-401 plastic signs are ideal for use in light duty-industrial, utility, commercial, and institutional environments
  • Durable non-reflective Aluminum (B-555) is a rigid aluminum panel with protected graphics designed for use in industrial, utility, commercial, and institutional environments
  • Self-Sticking Polyester (B-302) is an indoor/outdoor high performance pressure sensitive sign that is surface printed polyester with a polyester overlaminate
Biohazard indoor/outdoor signs are available in a variety of materials, allowing users to choose the material and size that best fits any application.