Brain-Heart Infusion Agar Plates, Teknova
Supplier: Teknova
B1010 B1010 B1010
100216-532PK 125.79 USD
100216-532 CA100216-532 77443-496
Brain-Heart Infusion Agar Plates, Teknova
Media for Microbiology Prepared Media for Microbiology
Brain Heart Infusion (BHI) Agar is a general-purpose medium suitable for the cultivation of a wide variety of organisms, including bacteria, yeasts and molds.
  • Sleeve of 20 plates
  • Sterile
Contains 3.7% Brain Heart Infusion Powder (37.0 g/L), 1.5% Agar (15.0 g/L)
Ambient shipping conditions. Store at room temperature.
Final pH of 7.4.