Carborundum Granules, 133B, Hengar
Supplier: Hengar
902100
43300-306EA 364.32 USD
43300-306
Carborundum Granules, 133B, Hengar
Boiling Stones
Hengar granules effectively prevent super-heating with its attendant dangerous explosive vaporization commonly known as "bumping".
  • Twelve mesh granule size
  • Hard, bluish black, iridescent silicon carbide granule
  • No reduction characteristics
Oxidizes slowly at 800 °C
Specific gravity 3.20