Thermostable RNase H

Supplier: Lucigen
H39500
76081-726EA 803.12 USD
76081-726
Thermostable RNase H
Enzymes
Specifically degrade the RNA in a DNA:RNA hybrid, without affecting DNA or unhybridized 

  • Optimal activity above 65°C and maintains activity as high as 95°C
  • Highly specific for RNA in a RNA:DNA hybrid and will not digest free RNA or DNA
  • Maximizes digestion sensitivity and selectivity while minimizing background due to nonspecific hybridization

Hybridase™ Thermostable RNase H specifically degrades the RNA in a DNA:RNA hybrid, without affecting DNA or unhybridized RNA

Applications include:
High-stringency hybrid selection
Diagnostic assays of specific target DNA sequences by isothermal probe amplification
Transcription-based amplification methods (e.g., NASBA® method)
High-stringency mapping of mRNA structure
Applications that require specific hydrolysis of the RNA in a DNA:RNA hybrid

In contrast to E. coli RNase H, which is rapidly inactivated at 55°C, Hybridase RNase H is active at high temperatures. It has optimal activity above 65°C and can be used at temperatures up to 95°C. The thermostability of the enzyme permits it to be used at temperatures that give the highest hybridization stringency for specific DNA:RNA heteroduplexes, maximizing sensitivity and selectivity while minimizing background due to nonspecific hybridization.

Certifications: For research use only. Not for human or diagnostic use.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR