Biox Multi-Well Culture Plates
Supplier: TradeWinds (TWD)
Biox
07-6006 07-6012 07-6096 07-6048 07-6024
76081-558CS 251.81 USD
76081-558 76081-564 76081-556 76081-560 76081-562
Biox Multi-Well Culture Plates
Microplates Cell Culture Plates
Cell Culture plates are sterile, constructed of prime virgin polystyrene, and a great value.
  • Prime virgin polysyrene
  • Individually wrapped packaging
  • Sterile
  • Cover design minimizes cross contamination
  • Thin bottom wall increases thermal consisency
  • Flat well bottom provides excellent clarity
TC treated plates come with a cover and feature flat bottoms for excellent clarity for microsopic examination of samples.
Certifications: ISO Certified: GB/T 19001-2008/ISO 9001:2008.