L-Carnosine, [3H] ≥97%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT994-250UCI MT994-1MCI
101974-074EA 2700.38 USD
101974-074 101974-076
L-Carnosine, [3H] ≥97%, MORpure™
L-Carnosine, [3H]
CAS Number: 2088050-69-3
UN: 2910
ADR: ,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR