Acetic acid, tetraethylammonium salt, [methyl-3H] 97, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT968-1MCI MT968-5MCI
101974-042EA 2726.54 USD
101974-042 101974-044
Acetic acid, tetraethylammonium salt, [methyl-3H] 97, MORpure™
Acetic acid, tetraethylammonium salt, [methyl-3H]
CAS Number: 340683-54-7
UN: 2910

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR