D-Alanine, [2,3-3H] ≥97%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT967-250UCI MT967-1MCI MT967-5MCI
101974-036EA 2700.38 USD
101974-036 101974-038 101974-040
D-Alanine, [2,3-3H] ≥97%, MORpure™
D-Alanine, [2,3-3H]
CAS Number: 313506-80-8
UN: 2910

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR