Water, [3H] ≥98% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT924B-1MCI
101973-970EA 588.46 USD
101973-970
Water, [3H] ≥98% (by HPLC), MORpure™
Water, [3H]
CAS Number: 13670-17-2
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR