L-Alanine, [2,3-3H] ≥96%

Supplier: MORAVEK BIOCHEMICALS MS

MT886-250UCI MT886-1MCI MT886-5MCI
101973-836EA 451.15 USD
101973-836 101973-838 101973-840
L-Alanine, [2,3-3H] ≥96%
L-Alanine, [2,3-3H]
CAS Number: 56877-49-7
UN: 2910
ADR: ,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR