2',3'-Dideoxycytidine, [5,6-3H] ≥97%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT812-250UCI MT812-500UCI MT812-1MCI
101973-670EA 2700.38 USD
101973-670 101973-672 101973-674
2',3'-Dideoxycytidine, [5,6-3H] ≥97%, MORpure™
2',3'-Dideoxycytidine, [5,6-3H]
CAS Number: 339532-23-9
UN: 2910

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR