Guanine-β-D-arabinofuranoside, [8-3H] ≥97% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT809-250UCI MT809-1MCI
101973-656EA 2700.38 USD
101973-656 101973-658
Guanine-β-D-arabinofuranoside, [8-3H] ≥97% (by HPLC), MORpure™
Guanine-β-D-arabinofuranoside, [8-3H]
CAS Number: 339531-49-6
UN: 2910
ADR: ,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR