Folic acid, diammonium salt, [3',5',7,9-3H] ≥97% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT783-50UCI MT783-250UCI MT783-500UCI MT783-1MCI
101973-592EA 2654.62 USD
101973-592 101973-594 101973-596 101973-598
Folic acid, diammonium salt, [3',5',7,9-3H] ≥97% (by HPLC), MORpure™
Folic acid, diammonium salt, [3',5',7,9-3H]
CAS Number: 339529-03-2
UN: 2910
ADR: ,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR