Adefovir, [adenine-2,8-3H] ≥97% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT703-250UCI MT703-1MCI
101973-450EA 2700.38 USD
101973-450 101973-452
Adefovir, [adenine-2,8-3H] ≥97% (by HPLC), MORpure™
Adefovir, [adenine-2,8-3H]
CAS Number: 339340-46-4
UN: 2910
ADR: ,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR