Thymidine, [methyl-3H] ≥97% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT6036-250UCI MT6036-1MCI MT6036-5MCI
101973-148EA 536.15 USD
101973-148 101973-150 101973-152
Thymidine, [methyl-3H] ≥97% (by HPLC), MORpure™
Thymidine, [methyl-3H]
CAS Number: 52497-68-4
UN: 2910
ADR: ,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR