D-Glucose, [2-3H(N)] ≥97% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT555W-250UCI MT555W-1MCI MT555W-5MCI
101973-076EA 503.46 USD
101973-076 101973-078 101973-080
D-Glucose, [2-3H(N)] ≥97% (by HPLC), MORpure™
D-Glucose, [2-3H(N)]
CAS Number: 2086328-04-1
UN: 2910
ADR: ,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR