Paclitaxel, [3H] ≥97%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT552-50UCI MT552-250UCI MT552-1MCI
101973-058EA 1601.92 USD
101973-058 101973-060 101973-062
Paclitaxel, [3H] ≥97%, MORpure™
Paclitaxel, [3H]
CAS Number: 145712-02-3
UN: 2910
ADR: ,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR