Cytidine, [6-3H] ≥97% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT1966-50UCI MT1966-250UCI
101972-890EA 1601.92 USD
101972-890 101972-892
Cytidine, [6-3H] ≥97% (by HPLC), MORpure™
Cytidine, [6-3H]
CAS Number: 2086327-35-5
UN: 2910
ADR: ,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR