Docetaxel
Supplier: MORAVEK BIOCHEMICALS MS
|
|
MT1914-50UCI
MT1914-250UCI
101972-778EA
1634.62
USD
101972-778
101972-780
Docetaxel
Docetaxel
Formula:
C₄₃H₅₃NO₁₄ |
MDL Number:
MFCD00800737 CAS Number: 114977-28-5 UN: 2910 ADR: , |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...