Phenytoin, [3H] ≥97% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT1862-250UCI
101972-658EA 2576.15 USD
101972-658
Phenytoin, [3H] ≥97% (by HPLC), MORpure™
Phenytoin, [3H]
CAS Number: 72670-37-2
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR