Ethanol, [3H] ≥96%

Supplier: MORAVEK BIOCHEMICALS MS

MT1766-1MCI MT1766-5MCI MT1766-25MCI
101972-582EA 2942.31 USD
101972-582 101972-584 101972-586
Ethanol, [3H] ≥96%
Ethanol, [3H]
CAS Number: 51628-31-0
UN: 2910

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR