Glycylsarcosine, [3H] ≥97% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT1545-50UCI MT1545-250UCI
101972-222EA 2556.54 USD
101972-222 101972-224
Glycylsarcosine, [3H] ≥97% (by HPLC), MORpure™
Glycylsarcosine, [3H]
CAS Number: 330795-61-4
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR