Benzyl alcohol, [7-14C] ≥98%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC478-250UCI MC478-1MCI
101971-992EA 5969.62 USD
101971-992 101971-994
Benzyl alcohol, [7-14C] ≥98%, MORpure™
Benzyl alcohol, [7-14C]
CAS Number: 13057-53-9
UN: 2910
ADR: ,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR