L-Tryptophan, [phenyl-14C(U)] ≥97%

Supplier: MORAVEK BIOCHEMICALS MS

MC2335-10UCI MC2335-50UCI
101971-434EA 1693.46 USD
101971-434 101971-436
L-Tryptophan, [phenyl-14C(U)] ≥97%
L-Tryptophan, [phenyl-14C(U)]
CAS Number: 2088208-92-6
UN: 2910

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR