Conjugated linoleic acid, [linoleic acid-1-14C] (mixture of isomers) ≥97% (by HPLC)

Supplier: MORAVEK BIOCHEMICALS MS

MC2323-50UCI
101971-404EA 5191.54 USD
101971-404
Conjugated linoleic acid, [linoleic acid-1-14C] (mixture of isomers) ≥97% (by HPLC)
Conjugated linoleic acid, [linoleic acid-1-14C] (mixture of isomers)
CAS Number: 313263-06-8
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR